siRNA Details
| Gene Symbol: Golga1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290649.1 | NP_001277578.1 | 18 | 2114 | ||
| NM_029793.2 | NP_084069.1 | 19 | 2189 | |||
| GenBank mRNA | AK028516.1 | - | 19 | 2109 | ||
| AK044987.1 | - | 18 | 2599 | |||
| BC037037.1 | - | 14 | 1525 | |||
| BC043090.1 | - | 19 | 2054 | 6418353 | ||
| BC053068.1 | - | 18 | 2042 | 6405595 | ||
| BC080733.1 | - | 18 | 2114 | 30110235 | ||

Golga1
Genomic Map