siRNA Details
| Gene Symbol: Arrdc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029799.1 | NP_084075.1 | 1 | 229 | ||
| GenBank mRNA | BC100496.1 | - | 1 | 229 | 30916615 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arrdc5| Gene Symbol: Arrdc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029799.1 | NP_084075.1 | 1 | 229 | ||
| GenBank mRNA | BC100496.1 | - | 1 | 229 | 30916615 | |