siRNA Details
| Gene Symbol: 2610028H24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029816.2 | NP_084092.2 | 5 | 569 | ||
| GenBank mRNA | AK011591.1 | - | 5 | 525 | ||
| AK161552.1 | - | 5 | 379 | |||
| BC051454.1 | - | 5 | 506 | 6308081 | ||
| BQ714071.1 | - | 5 | 588 | |||

2610028H24Rik
Genomic Map