siRNA Details
| Gene Symbol: 2610028H24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029816.2 | NP_084092.2 | 1 | 226 | ||
| GenBank mRNA | AK011591.1 | - | 1 | 182 | ||
| BC051454.1 | - | 1 | 163 | 6308081 | ||
| BQ714071.1 | - | 1 | 245 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2610028H24Rik| Gene Symbol: 2610028H24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029816.2 | NP_084092.2 | 1 | 226 | ||
| GenBank mRNA | AK011591.1 | - | 1 | 182 | ||
| BC051454.1 | - | 1 | 163 | 6308081 | ||
| BQ714071.1 | - | 1 | 245 | |||