siRNA Details
| Gene Symbol: Scfd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014070.1 | - | 16, 17 | 1410 | ||
| AK044943.1 | - | 16, 17 | 1411 | |||
| BC027793.1 | - | 16, 17 | 1429 | 5151118 | ||
| CO044480.1 | - | 9, 10 | 697 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scfd1| Gene Symbol: Scfd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014070.1 | - | 16, 17 | 1410 | ||
| AK044943.1 | - | 16, 17 | 1411 | |||
| BC027793.1 | - | 16, 17 | 1429 | 5151118 | ||
| CO044480.1 | - | 9, 10 | 697 | |||