siRNA Details
| Gene Symbol: Scfd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014070.1 | - | 4 | 271 | ||
| AK041028.1 | - | 4 | 270 | |||
| AK044943.1 | - | 4 | 272 | |||
| AK049361.1 | - | 4 | 271 | |||
| AK050504.1 | - | 4 | 270 | |||
| AK129238.1 | - | 4 | 239 | |||
| BC027793.1 | - | 4 | 290 | 5151118 | ||

Scfd1
Genomic Map