siRNA Details
Gene Symbol: Ccnc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290420.1 | NP_001277349.1 | 12 | 1292 | ||
GenBank mRNA | AK135472.1 | - | 12 | 1288 |
Gene Symbol: Tstd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029840.1 | NP_084116.1 | 2 | 216 | ||
GenBank mRNA | AI287097.1 | - | 2 | 216 | ||
AK011611.1 | - | 2 | 131 | |||
AK154853.1 | - | 2 | 203 | |||
AK218953.1 | - | 1 | 56 |
