siRNA Details
| Gene Symbol: Arid2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175251.4 | NP_780460.3 | 18 | 5275 | ||
| GenBank mRNA | AK014481.1 | - | 5 | 3240 | ||
| AK034964.1 | - | 4 | 1436 | |||
| AK077022.1 | - | 3 | 349 | |||
| AK077366.1 | - | 4 | 2037 | |||
| AK173201.1 | - | 4 | 3102 | |||
| BC027180.1 | - | 4 | 2174 | 4021271 | ||

Arid2
Genomic Map