siRNA Details
| Gene Symbol: Ints7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK034856.1 | - | 11 | 1385 | ||
| AK040344.1 | - | 9 | 1163 | |||
| AK052103.1 | - | 9 | 1113 | |||
| AK085336.1 | - | 10 | 1259 | |||
| AK085362.1 | - | 11 | 1424 | |||
| AK086462.1 | - | 11 | 1417 | |||
| BC054120.1 | - | 11 | 1370 | 6506658 | ||

Ints7
Genomic Map