siRNA Details
Gene Symbol: Klhl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178633.3 | NP_848748.2 | 792 | |||
GenBank mRNA | AK032116.1 | - | 340 | |||
AK167903.1 | - | 583 | ||||
AK220570.1 | - | 792 | ||||
BC031142.1 | - | 663 | 4972965 | |||
BC031144.1 | - | 686 | 4973325 |
