siRNA Details
| Gene Symbol: Mtmr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023858.3 | NP_076347.3 | 14 | 1758 | ||
| GenBank mRNA | AK133476.1 | - | 14 | 1758 | ||
| AK133889.1 | - | 13 | 1526 | |||
| AK191371.1 | - | 2 | 130 | |||
| AK220369.1 | - | 7 | 7354 | |||
| AY055832.1 | - | 14 | 2529 | |||
| BC027013.1 | - | 5 | 1335 | |||
| BC063050.1 | - | 15 | 1847 | 5716297 | ||

Mtmr2
Genomic Map