siRNA Details
| Gene Symbol: Mtmr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023858.3 | NP_076347.3 | 3 | 344 | ||
| GenBank mRNA | AF073880.1 | - | 3 | 210 | ||
| AK020065.1 | - | 4 | 416 | |||
| AK133476.1 | - | 3 | 344 | |||
| AK133889.1 | - | 2 | 111 | |||
| AK176943.1 | - | 3 | 161 | |||
| AY055832.1 | - | 3 | 1115 | |||
| BC063050.1 | - | 4 | 433 | 5716297 | ||

Mtmr2
Genomic Map