siRNA Details
| Gene Symbol: Zfp142 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029888.2 | NP_084164.1 | 3 | 970 | ||
| GenBank mRNA | AB093218.1 | - | 3 | 1257 | ||
| AK132550.1 | - | 3 | 971 | |||
| BC060118.1 | - | 3 | 906 | 6406631 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp142| Gene Symbol: Zfp142 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029888.2 | NP_084164.1 | 3 | 970 | ||
| GenBank mRNA | AB093218.1 | - | 3 | 1257 | ||
| AK132550.1 | - | 3 | 971 | |||
| BC060118.1 | - | 3 | 906 | 6406631 | ||