siRNA Details
| Gene Symbol: Zfp142 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029888.2 | NP_084164.1 | 8 | 5218 | ||
| GenBank mRNA | AB093218.1 | - | 8 | 5505 | ||
| AF332091.1 | - | 7 | 4506 | |||
| AF332092.1 | - | 7 | 4506 | |||
| AK052201.1 | - | 2 | 2159 | |||
| AK052359.1 | - | 2 | 1198 | |||
| AK132550.1 | - | 8 | 5219 | |||
| BC060118.1 | - | 8 | 5152 | 6406631 | ||

Zfp142
Genomic Map