siRNA Details
| Gene Symbol: Heg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175256.5 | NP_780465.4 | 15 | 3346 | ||
| GenBank mRNA | AK043598.1 | - | 4 | 1584 | ||
| BC072651.1 | - | 7 | 656 | |||
| BC076596.1 | - | 13 | 2625 | 30645937 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Heg1| Gene Symbol: Heg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175256.5 | NP_780465.4 | 15 | 3346 | ||
| GenBank mRNA | AK043598.1 | - | 4 | 1584 | ||
| BC072651.1 | - | 7 | 656 | |||
| BC076596.1 | - | 13 | 2625 | 30645937 | ||