siRNA Details
Gene Symbol: Tmem116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161626.1 | NP_001155098.1 | 10 | 885 | ||
NM_001161627.1 | NP_001155099.1 | 11 | 990 | |||
NM_029912.2 | NP_084188.1 | 10 | 923 | |||
GenBank mRNA | AK021099.1 | - | 10 | 929 | ||
BC019151.1 | - | 10 | 880 | 5064772 |
