siRNA Details
| Gene Symbol: Tmem116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161626.1 | NP_001155098.1 | 7, 8 | 600 | ||
| NM_001161627.1 | NP_001155099.1 | 8, 9 | 705 | |||
| NM_029912.2 | NP_084188.1 | 7, 8 | 638 | |||
| GenBank mRNA | AK015103.1 | - | 9, 10 | 855 | ||
| AK021099.1 | - | 7, 8 | 644 | |||
| AK039773.1 | - | 1149 | ||||
| BC019151.1 | - | 7, 8 | 595 | 5064772 | ||
| CK127968.1 | - | 8, 9 | 705 | |||
| DV658154.1 | - | 5, 6 | 439 | |||

Tmem116
Genomic Map