siRNA Details
Gene Symbol: Kidins220 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081378.1 | NP_001074847.1 | 25 | 3603 | ||
GenBank mRNA | AK039537.1 | - | 4 | 495 | ||
AK083260.1 | - | 19 | 2927 | |||
AK122478.1 | - | 22 | 3194 | |||
KJ812113.1 | - | 2 | 247 | |||
KJ812114.1 | - | 2 | 247 | |||
KJ812115.1 | - | 2 | 247 | |||
KJ812116.1 | - | 2 | 247 |
