siRNA Details
| Gene Symbol: Kidins220 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081378.1 | NP_001074847.1 | 22 | 3104 | ||
| GenBank mRNA | AK083260.1 | - | 16 | 2428 | ||
| BC044772.2 | - | 17 | 2460 | |||
| BC046467.1 | - | 17 | 2460 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kidins220| Gene Symbol: Kidins220 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081378.1 | NP_001074847.1 | 22 | 3104 | ||
| GenBank mRNA | AK083260.1 | - | 16 | 2428 | ||
| BC044772.2 | - | 17 | 2460 | |||
| BC046467.1 | - | 17 | 2460 | |||