siRNA Details
| Gene Symbol: Tcaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029930.2 | NP_084206.1 | 3 | 1666 | ||
| GenBank mRNA | AK014379.1 | - | 3 | 1195 | ||
| AK053509.1 | - | 3 | 1667 | |||
| AK076242.1 | - | 3 | 1612 | |||
| AK083917.1 | - | 3 | 1614 | |||
| AK132148.1 | - | 3 | 1634 | |||
| AK148593.1 | - | 3 | 1645 | |||
| AK173013.1 | - | 3 | 1648 | |||
| BC031718.1 | - | 3 | 1694 | 3156550 | ||

Tcaf1
Genomic Map