siRNA Details
| Gene Symbol: Tcaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029930.2 | NP_084206.1 | 2 | 640 | ||
| GenBank mRNA | AK020704.1 | - | 3 | 736 | ||
| AK053509.1 | - | 2 | 641 | |||
| AK076242.1 | - | 2 | 586 | |||
| AK083917.1 | - | 2 | 588 | |||
| AK132148.1 | - | 2 | 608 | |||
| AK148593.1 | - | 2 | 619 | |||
| AK173013.1 | - | 2 | 622 | |||

Tcaf1
Genomic Map