siRNA Details
| Gene Symbol: Bcl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029933.4 | NP_084209.3 | 8 | 2320 | ||
| GenBank mRNA | AK084676.1 | - | 1 | 247 | ||
| AK147659.1 | - | 8 | 2320 | |||
| AY296061.1 | - | 5 | 1836 | |||
| BC137850.1 | - | 5 | 1808 | 8860869 | ||

Bcl9
Genomic Map