siRNA Details
| Gene Symbol: Myh10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175260.2 | NP_780469.1 | 12 | 1650 | ||
| GenBank mRNA | AK029236.1 | - | 12 | 1457 | ||
| AK045066.1 | - | 12 | 1452 | |||
| AK045074.1 | - | 12 | 1452 | |||
| AK052435.1 | - | 12 | 1455 | |||
| AK122578.1 | - | 10 | 922 | |||
| AK133946.1 | - | 11 | 1416 | |||
| AK147567.1 | - | 12 | 1629 | |||
| AK163604.1 | - | 12 | 1466 | |||
| BC089011.1 | - | 12 | 1426 | 30620877 | ||

Myh10
Genomic Map