siRNA Details
| Gene Symbol: Myh10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175260.2 | NP_780469.1 | 16 | 2217 | ||
| GenBank mRNA | AK045066.1 | - | 16 | 2019 | ||
| AK045074.1 | - | 16 | 2019 | |||
| AK052435.1 | - | 16 | 2022 | |||
| AK122578.1 | - | 14 | 1489 | |||
| AK147567.1 | - | 16 | 2196 | |||
| AK163604.1 | - | 16 | 2033 | |||
| BC089011.1 | - | 16 | 1993 | 30620877 | ||

Myh10
Genomic Map