siRNA Details
| Gene Symbol: Myh10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175260.2 | NP_780469.1 | 2 | 383 | ||
| GenBank mRNA | AK029236.1 | - | 2 | 190 | ||
| AK045066.1 | - | 2 | 185 | |||
| AK045074.1 | - | 2 | 185 | |||
| AK047340.1 | - | 2 | 218 | |||
| AK052435.1 | - | 2 | 188 | |||
| AK147567.1 | - | 2 | 362 | |||
| AK163604.1 | - | 2 | 199 | |||
| BC089011.1 | - | 2 | 159 | 30620877 | ||
| CJ100063.1 | - | 2 | 383 | |||

Myh10
Genomic Map