siRNA Details
| Gene Symbol: Usp45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290425.1 | NP_001277354.1 | 14, 15 | 1991 | ||
| NM_152825.2 | NP_690038.1 | 15, 16 | 2135 | |||
| GenBank mRNA | AK019756.1 | - | 16, 17 | 2250 | ||
| AK036903.1 | - | 2, 3 | 551 | |||
| AK088073.1 | - | 14, 15 | 1974 | |||
| BC027768.1 | - | 15, 16 | 2118 | 5354502 | ||
| BM227454.2 | - | 3, 4 | 501 | |||

Usp45
Genomic Map