siRNA Details
| Gene Symbol: Prelid2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029942.1 | NP_084218.1 | 4, 5 | 390 | ||
| GenBank mRNA | AK021193.1 | - | 4, 5 | 416 | ||
| BC119251.1 | - | 4, 5 | 390 | 8734000 | ||
| BC120711.1 | - | 4, 5 | 390 | 40129634 | ||
| BC145035.1 | - | 4, 5 | 390 | 9053583 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001271333, XM_008772173, XM_008772174.

Prelid2
Genomic Map