siRNA Details
| Gene Symbol: Prelid2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029942.1 | NP_084218.1 | 3 | 193 | ||
| GenBank mRNA | AK021193.1 | - | 3 | 219 | ||
| BC119251.1 | - | 3 | 193 | 8734000 | ||
| BC120711.1 | - | 3 | 193 | 40129634 | ||
| BC145035.1 | - | 3 | 193 | 9053583 | ||

Prelid2
Genomic Map