siRNA Details
| Gene Symbol: Sphkap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172430.3 | NP_766018.3 | 7 | 3579 | ||
| GenBank mRNA | AK019735.1 | - | 1 | 700 | ||
| AK038638.1 | - | 8 | 3624 | |||
| AK122538.1 | - | 1 | 2529 | |||
| BC042654.1 | - | 1 | 434 | |||
| BC060165.1 | - | 1 | 712 | |||
| BC060217.1 | - | 1 | 1085 | |||
| BC069832.1 | - | 7 | 3545 | 30431760 | ||

Sphkap
Genomic Map