siRNA Details
| Gene Symbol: Trat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198297.3 | NP_938039.2 | 3 | 176 | ||
| GenBank mRNA | AJ297969.1 | - | 3 | 194 | ||
| AK030807.1 | - | 3 | 359 | |||
| AK138735.1 | - | 242 | ||||
| BC104351.1 | - | 3 | 176 | 40049558 | ||

Trat1
Genomic Map