siRNA Details
| Gene Symbol: Trat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198297.3 | NP_938039.2 | 2 | 100 | ||
| GenBank mRNA | AJ297969.1 | - | 2 | 118 | ||
| AK030807.1 | - | 2 | 283 | |||
| AK138735.1 | - | 166 | ||||
| AK163002.1 | - | 1 | 214 | |||
| BC104350.1 | - | 2 | 100 | 40049557 | ||
| BC104351.1 | - | 2 | 100 | 40049558 | ||

Trat1
Genomic Map