siRNA Details
| Gene Symbol: Arhgef38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029953.1 | NP_084229.1 | 3 | 492 | ||
| GenBank mRNA | BC049886.1 | - | 3 | 492 | 5387778 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arhgef38| Gene Symbol: Arhgef38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029953.1 | NP_084229.1 | 3 | 492 | ||
| GenBank mRNA | BC049886.1 | - | 3 | 492 | 5387778 | |