siRNA Details
| Gene Symbol: Arhgef38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029953.1 | NP_084229.1 | 1 | 178 | ||
| GenBank mRNA | AK020284.1 | - | 1 | 239 | ||
| BC049886.1 | - | 1 | 178 | 5387778 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arhgef38| Gene Symbol: Arhgef38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029953.1 | NP_084229.1 | 1 | 178 | ||
| GenBank mRNA | AK020284.1 | - | 1 | 239 | ||
| BC049886.1 | - | 1 | 178 | 5387778 | ||