siRNA Details
| Gene Symbol: Elp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023876.3 | NP_076365.2 | 7, 8 | 926 | ||
| GenBank mRNA | AJ276004.1 | - | 7, 8 | 925 | ||
| AK034040.1 | - | 7, 8 | 931 | |||
| AK034529.1 | - | 7, 8 | 932 | |||
| AK037104.1 | - | 7, 8 | 950 | |||
| AK039447.1 | - | 7, 8 | 938 | |||
| AK039758.1 | - | 7, 8 | 927 | |||
| AK079069.1 | - | 7, 8 | 941 | |||
| AK168964.1 | - | 7, 8 | 990 | |||

Elp4
Genomic Map