siRNA Details
| Gene Symbol: Adamtsl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029981.1 | NP_084257.1 | 19 | 2984 | ||
| GenBank mRNA | AB096928.1 | - | 19 | 2984 | ||
| AK020831.1 | - | 3 | 446 | |||
| AK156991.1 | - | 19 | 2987 | |||
| AK172978.1 | - | 7 | 5390 | |||
| AK181027.1 | - | 154 | ||||
| BC128568.1 | - | 19 | 2936 | 40097415 | ||
| BC129984.1 | - | 19 | 2936 | 40097418 | ||

Adamtsl2
Genomic Map