siRNA Details
| Gene Symbol: Krba1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133922.3 | NP_598683.3 | 18 | 3212 | ||
| GenBank mRNA | AB041543.1 | - | 16 | 2844 | ||
| AK029235.1 | - | 8 | 2210 | |||
| AK038090.1 | - | 4 | 862 | |||
| AK040525.1 | - | 18 | 3214 | |||
| AK128931.1 | - | 18 | 3163 | |||
| AK162934.1 | - | 18 | 2920 | |||
| AK173281.1 | - | 18 | 4916 | |||
| BC066816.1 | - | 18 | 3132 | 30084911 | ||

Krba1
Genomic Map