siRNA Details
| Gene Symbol: Mlana | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029993.1 | NP_084269.1 | 5 | 353 | ||
| GenBank mRNA | AK020928.1 | - | 5 | 372 | ||
| BC111114.1 | - | 5 | 353 | 40053172 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mlana| Gene Symbol: Mlana | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029993.1 | NP_084269.1 | 5 | 353 | ||
| GenBank mRNA | AK020928.1 | - | 5 | 372 | ||
| BC111114.1 | - | 5 | 353 | 40053172 | ||