siRNA Details
Gene Symbol: Fam53b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175268.4 | NP_780477.2 | 4 | 413 | ||
NM_212473.1 | NP_997638.1 | 4 | 476 | |||
GenBank mRNA | AK020830.1 | - | 4 | 414 | ||
AK031576.1 | - | 3 | 257 | |||
AK051299.1 | - | 4 | 477 | |||
AK147350.1 | - | 4 | 809 | |||
AK147725.1 | - | 4 | 605 | |||
AK156629.1 | - | 4 | 376 | |||
AK187644.1 | - | 3 | 229 | |||
AK195672.1 | - | 3 | 181 | |||
BC035523.1 | - | 4 | 408 | 5400742 |
