siRNA Details
| Gene Symbol: Rpgrip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168515.1 | NP_001161987.1 | 7, 8 | 893 | ||
| NM_023879.3 | NP_076368.1 | 7, 8 | 893 | |||
| GenBank mRNA | AK086397.1 | - | 4, 5 | 587 | ||
| AY008297.1 | - | 7, 8 | 875 | |||
| AY914560.1 | - | 7, 8 | 869 | |||
| BC016092.1 | - | 7, 8 | 855 | |||
| BC081545.1 | - | 7, 8 | 893 | |||
| BC086692.1 | - | 7, 8 | 857 | 30682813 | ||
| HM067441.1 | - | 6, 7 | 797 | |||

Rpgrip1
Genomic Map