siRNA Details
| Gene Symbol: Rpgrip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168515.1 | NP_001161987.1 | 13 | 1566 | ||
| NM_023879.3 | NP_076368.1 | 13 | 1566 | |||
| GenBank mRNA | AK015701.1 | - | 3 | 565 | ||
| AK029955.1 | - | 3 | 514 | |||
| AY008297.1 | - | 13 | 1548 | |||
| AY914560.1 | - | 13 | 1539 | |||
| BC016092.1 | - | 13 | 1525 | |||
| BC081545.1 | - | 13 | 1561 | |||
| BC086692.1 | - | 13 | 1530 | 30682813 | ||
| HM067441.1 | - | 12 | 1470 | |||

Rpgrip1
Genomic Map