siRNA Details
| Gene Symbol: Hook1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030014.2 | NP_084290.1 | 19 | 1879 | ||
| GenBank mRNA | AF487912.1 | - | 19 | 1961 | ||
| BC061688.1 | - | 19 | 1879 | 5364426 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hook1| Gene Symbol: Hook1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030014.2 | NP_084290.1 | 19 | 1879 | ||
| GenBank mRNA | AF487912.1 | - | 19 | 1961 | ||
| BC061688.1 | - | 19 | 1879 | 5364426 | ||