siRNA Details
Gene Symbol: Ccdc150 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030025.2 | NP_084301.2 | 25 | 3033 | ||
GenBank mRNA | AK029994.1 | - | 25 | 3033 | ||
BC145597.1 | - | 27 | 3123 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ccdc150 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030025.2 | NP_084301.2 | 25 | 3033 | ||
GenBank mRNA | AK029994.1 | - | 25 | 3033 | ||
BC145597.1 | - | 27 | 3123 |