siRNA Details
| Gene Symbol: Sowahb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175270.4 | NP_780479.1 | 1 | 1354 | ||
| GenBank mRNA | AK033423.1 | - | 1 | 1284 | ||
| AK033641.1 | - | 1 | 817 | |||
| AK137429.1 | - | 1 | 1355 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sowahb| Gene Symbol: Sowahb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175270.4 | NP_780479.1 | 1 | 1354 | ||
| GenBank mRNA | AK033423.1 | - | 1 | 1284 | ||
| AK033641.1 | - | 1 | 817 | |||
| AK137429.1 | - | 1 | 1355 | |||