siRNA Details
| Gene Symbol: Spint4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030058.2 | NP_084334.1 | 2 | 272 | ||
| GenBank mRNA | AK020315.1 | - | 2 | 245 | ||
| AY226988.1 | - | 2 | 272 | |||
| BC119299.1 | - | 2 | 242 | 8734048 | ||
| BC119301.1 | - | 2 | 242 | 8734050 | ||

Spint4
Genomic Map