siRNA Details
| Gene Symbol: Armcx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166377.1 | NP_001159849.1 | 6 | 1098 | ||
| NM_001166378.1 | NP_001159850.1 | 4 | 797 | |||
| NM_001166379.1 | NP_001159851.1 | 4 | 913 | |||
| NM_001166380.1 | NP_001159852.1 | 5 | 889 | |||
| NM_030066.3 | NP_084342.1 | 6 | 982 | |||
| GenBank mRNA | AK019405.1 | - | 6 | 909 | ||
| AK147002.1 | - | 6 | 1049 | |||
| BC021410.1 | - | 5 | 817 | 5007058 | ||
| BC026488.1 | - | 4 | 881 | 4504219 | ||
| BC068228.1 | - | 6 | 931 | 6413546 | ||
| CB209449.1 | - | 4 | 745 | |||

Armcx1
Genomic Map