siRNA Details
| Gene Symbol: Lrrc9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142728.1 | NP_001136200.1 | 18 | 2586 | ||
| NM_030070.3 | NP_084346.2 | 18 | 2589 | |||
| GenBank mRNA | AK019545.1 | - | 18 | 2522 | ||
| AK029802.1 | - | 18 | 2587 | |||

Lrrc9
Genomic Map