siRNA Details
| Gene Symbol: Rbsn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030081.2 | NP_084357.2 | 12 | 1967 | ||
| GenBank mRNA | BC031135.1 | - | 11 | 1798 | 4971294 | |
| BC049106.1 | - | 12 | 1967 | 6412839 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rbsn| Gene Symbol: Rbsn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030081.2 | NP_084357.2 | 12 | 1967 | ||
| GenBank mRNA | BC031135.1 | - | 11 | 1798 | 4971294 | |
| BC049106.1 | - | 12 | 1967 | 6412839 | ||