siRNA Details
| Gene Symbol: Ndufv3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083891.1 | NP_001077360.1 | 2 | 185 | ||
| NM_030087.2 | NP_084363.2 | 2 | 185 | |||
| GenBank mRNA | AK018779.1 | - | 2 | 150 | ||
| AK077433.1 | - | 2 | 126 | |||
| AK154191.1 | - | 2 | 118 | |||
| AK154477.1 | - | 2 | 164 | |||
| AK160289.1 | - | 2 | 116 | |||
| BC013640.1 | - | 2 | 142 | 4188234 | ||
| BU555486.1 | - | 2 | 188 | |||
| BY064612.1 | - | 2 | 153 | |||

Ndufv3
Genomic Map