siRNA Details
| Gene Symbol: 2210407C18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144544.2 | NP_653127.2 | 4 | 375 | ||
| GenBank mRNA | AA124355.1 | - | 4 | 361 | ||
| AA434919.1 | - | 2 | 181 | |||
| AY148487.1 | - | 3 | 281 | |||
| BC019553.1 | - | 4 | 353 | 4456658 | ||
| BY097425.1 | - | 4 | 376 | |||

2210407C18Rik
Genomic Map