siRNA Details
| Gene Symbol: Helz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198298.1 | NP_938040.1 | 30 | 5053 | ||
| GenBank mRNA | AK033094.1 | - | 14 | 2708 | ||
| BC060114.1 | - | 4 | 345 | |||
| BC076626.1 | - | 30 | 5053 | 30640711 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Helz| Gene Symbol: Helz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198298.1 | NP_938040.1 | 30 | 5053 | ||
| GenBank mRNA | AK033094.1 | - | 14 | 2708 | ||
| BC060114.1 | - | 4 | 345 | |||
| BC076626.1 | - | 30 | 5053 | 30640711 | ||